Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UACA Antibody, Novus Biologicals™
SDP

Catalog No. NB10061622 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10061622 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10061622 Supplier Novus Biologicals Supplier No. NB10061622
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UACA Polyclonal specifically detects UACA in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen UACA
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot (Negative)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 ug/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_060473.2
Gene Alias FLJ10128, KIAA1561MGC141969, MGC141967, nuclear membrane binding protein, NUCLING, uveal autoantigen with coiled coil domains and ankyrin repeats, uveal autoantigen with coiled-coil domains and ankyrin repeats
Gene Symbols UACA
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 675 and 725 of human uveal autoantigen with coiled-coil domains and ankyrin repeats using the numbering given in entry NP_060473.2 (GeneID 55075).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 55075
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.