Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UAP56 Antibody (OTI2C5), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP274727R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274727R 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274727R Supplier Novus Biologicals Supplier No. NBP274727R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UAP56 Monoclonal specifically detects UAP56 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen UAP56
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C5
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias 56 kDa U2AF65-associated protein, ATP-dependent RNA helicase p47, DEAD (Asp-Glu-Ala-Asp) box polypeptide 39B, DEAD box protein UAP56, EC 3.6.4.13, HLA-B associated transcript 1, HLA-B-associated transcript 1 protein, spliceosome RNA helicase BAT1, UAP56D6S81EBAT1nuclear RNA helicase (DEAD family)
Gene Symbols DDX39B
Host Species Mouse
Immunogen Full length recombinant protein of human UAP56 (NP_542165) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Proteases & Other Enzymes
Primary or Secondary Primary
Gene ID (Entrez) 7919.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.