Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ub-K63 Antibody (HWA4C4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP20199A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP20199A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP20199A350 Supplier Novus Biologicals Supplier No. NBP200199AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ub-K63
Applications Western Blot
Classification Monoclonal
Clone HWA4C4
Conjugate Alexa Fluor 350
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Host Species Mouse
Immunogen The immunogen for this antibody is Ub-K63.
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.