Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ub-K63 Antibody (HWA4C4), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP200199IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP200199IR 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP200199IR Supplier Novus Biologicals Supplier No. NBP200199IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ub-K63 Monoclonal specifically detects Ub-K63 in Human samples. It is validated for Western Blot.

Specifications

Antigen Ub-K63
Applications Western Blot
Classification Monoclonal
Clone HWA4C4
Conjugate DyLight 755
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Host Species Mouse
Immunogen The immunogen for this antibody is Ub-K63.
Purification Method Protein A or G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity HWA4C4 reacts specifically with the isopeptide linkage region of lysine-63-linked polyubiquitin. It does not react with other forms of ubiquitin including ubiquitin monomer and lysine-48-linked chains.
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.