Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBA3 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NBP275926 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP275926 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP275926 Supplier Novus Biologicals Supplier No. NBP275926

Rabbit Polyclonal Antibody

UBA3 Polyclonal specifically detects UBA3 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen UBA3
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500, Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:50-1:200
Formulation PBS (pH7.4), 0.2% BSA, 50% Glycerol with 0.05% Sodium Azide
Gene Alias DKFZp566J164, EC 6.3.2, EC 6.3.2.-, hUba3, MGC22384, NEDD8-activating enzyme E1 catalytic subunit, NEDD8-activating enzyme E1C, Nedd8-activating enzyme hUba3, UBA3, ubiquitin-activating enzyme E1 homolog, UBE1C, Ubiquitin-activating enzyme 3, Ubiquitin-activating enzyme E1C, ubiquitin-activating enzyme E1C (homologous to yeast UBA3), ubiquitin-like modifier activating enzyme 3, Ubiquitin-like modifier-activating enzyme 3, yeast)
Gene Symbols UBA3
Host Species Rabbit
Immunogen Recombinant protein within Human UBA3 aa 220-424/ 463. (SwissProt: Q8TBC4 Human; SwissProt: Q8C878 Mouse; SwissProt: Q99MI7 Rat)
Molecular Weight of Antigen 52 kDa
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 9039.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.