Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ UBAP2L Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA572885
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
PIPA572885 100 μg
1 options

Catalog No. PIPA572885

Supplier: Invitrogen™ PA572885

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBAP2L Polyclonal Antibody for Western Blot, ICC/IF, ELISA

NICE4, also known as UBAP2L (Ubiquitin-associated protein 2-like) orKIAA0144, is a 1,087 amino acid protein that is ubiquitously expressed. Phosphorylated upon DNA damage, NICE4 contains one UBA domain and is expressed as 4 isoforms produced by alternative splicing events. The gene that encodes NICE4 maps to human chromosome 1. Chromosome 1 is the largest human chromosome spanning about 260 million base pairs and making up 8% of the human genome. There are about 3,000 genes on chromosome 1, and considering the great number of genes there are also a large number of diseases associated with chromosome 1. Notably, the rare aging disease Hutchinson-Gilford progeria is associated with the LMNA gene which encodes Lamin A. When defective, the LMNA gene product can build up in the nucleus and cause characteristic nuclear blebs. The mechanism of rapidly enhanced aging is unclear and is a topic of continuing exploration. The MUTYH gene is located on chromosome 1 and is partially responsible for familial adenomatous polyposis. Stickler syndrome, Parkinsons, Gaucher disease and Usher syndrome are also associated with chromosome 1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen UBAP2L
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene UBAP2L
Gene Accession No. Q14157, Q80X50
Gene Alias 3110083O19Rik; 4932431F02Rik; A430103N23Rik; Atp8b2; Atpase, class I, type 8B, member 2; C77168; EP1; KIAA0144; mKIAA0144; NICE4; NICE-4; Nice-4 protein homolog; Protein NICE-4; Ubap2l; ubiquitin associated protein 2 like; ubiquitin associated protein 2-like; ubiquitin-associated protein 2-like
Gene Symbols UBAP2L
Host Species Rabbit
Immunogen UBAP2L antibody for a 19 amino acid peptide from near the amino terminus of human UBAP2L. The immunogen is within amino acids 200 - 250 of UBAP2L.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 74383, 9898
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.