Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH10/UBE2C Antibody (CPTC-UBE2C-1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB378588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB378588 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB378588 Supplier Novus Biologicals Supplier No. NBP308829AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UbcH10/UBE2C Monoclonal antibody specifically detects UbcH10/UBE2C in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)

Specifications

Antigen UbcH10/UBE2C
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone CPTC-UBE2C-1
Conjugate Alexa Fluor 750
Formulation 50mM Sodium Borate
Gene Alias cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C
Host Species Mouse
Immunogen Recombinant human full-length UbcH10/UBE2C protein (Uniprot: O00762)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 11065
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.