Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH10/UBE2C Antibody, Novus Biologicals™
SDP

Catalog No. NB1001084 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001084 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001084 Supplier Novus Biologicals Supplier No. NB1001084
Add to Cart
Add to Cart

Goat Polyclonal Antibody

UbcH10/UBE2C Polyclonal specifically detects UbcH10/UBE2C in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot.

Specifications

Antigen UbcH10/UBE2C
Applications Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Peptide ELISA, Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Flow Cytometry 10 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunoprecipitation, Peptide ELISA Detection limit 1:32000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. NP_008950.
Gene Alias cyclin-selective ubiquitin carrier protein, dJ447F3.2, EC 6.3.2.19, UbcH10, UBCH10mitotic-specific ubiquitin-conjugating enzyme, Ubiquitin carrier protein C, ubiquitin carrier protein E2-C, ubiquitin-conjugating enzyme E2 C, ubiquitin-conjugating enzyme E2C, Ubiquitin-protein ligase C
Gene Symbols UBE2C
Host Species Goat
Immunogen Peptide with sequence C-QETYSKQVTSQEP corresponding to C-Terminus according to NP_008950.1, NP_861515.1, NP_861516.1, NP_861517.1, NP_861518.1.
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Mitotic Regulators
Primary or Secondary Primary
Gene ID (Entrez) 11065
Test Specificity This antibody is expected to recognize all reported isoforms. Variants NP_861517.1 and NP_861518.1 encode the same isoform.
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.