Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH2/UBE2H Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB064121 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB064121 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB064121 Supplier Novus Biologicals Supplier No. NBP300048AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UbcH2/UBE2H Polyclonal specifically detects UbcH2/UBE2H in Human samples. It is validated for Western Blot, ELISA.

Specifications

Antigen UbcH2/UBE2H
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, UBC8E2-20K, UBCH, UBCH2, Ubiquitin carrier protein H, ubiquitin-conjugating enzyme E2 H, Ubiquitin-conjugating enzyme E2-20K, ubiquitin-conjugating enzyme E2H (homologous to yeast UBC8), ubiquitin-conjugating enzyme E2H (UBC8 homolog, yeast), Ubiquitin-protein ligase H
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant UbcH2/UBE2H (P62256; Met1-Leu183).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7328
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.