Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH5b/UBE2D2 Antibody (OTI2C2), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP274732FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274732FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274732FR Supplier Novus Biologicals Supplier No. NBP274732FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UbcH5b/UBE2D2 Monoclonal specifically detects UbcH5b/UBE2D2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen UbcH5b/UBE2D2
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI2C2
Conjugate DyLight 680
Dilution Western Blot, Immunohistochemistry
Formulation PBS with 0.05% Sodium Azide
Gene Alias E2(17)KB2, EC 6.3.2.19, PUBC1, UBC4/5, UBC4ubiquitin-conjugating enzyme E2D 2 (homologous to yeast UBC4/5), UBC5B, UBCH4, UbcH5B, Ubiquitin carrier protein D2, ubiquitin-conjugating enzyme E2 D2, Ubiquitin-conjugating enzyme E2(17)KB 2, Ubiquitin-conjugating enzyme E2-17 kDa 2, ubiquitin-conjugating enzyme E2D 2 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase D2
Gene Symbols UBE2D2
Host Species Mouse
Immunogen Full length recombinant protein of human UbcH5b/UBE2D2 (NP_003330) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Cancer, Hypoxia
Primary or Secondary Primary
Gene ID (Entrez) 7322.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.