Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH7/UBE2L3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220786 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220786 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220786 Supplier Novus Biologicals Supplier No. NBP220786
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UbcH7/UBE2L3 Polyclonal specifically detects UbcH7/UBE2L3 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UbcH7/UBE2L3
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 0.78mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, Immunocytochemistry/Immunofluorescence 1:100-1:1000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. P68036
Gene Alias E2-F1, EC 6.3.2.19, L-UBC, UBCE7, UbcH7, UBCH7UbcM4, Ubiquitin carrier protein L3, ubiquitin-conjugating enzyme E2 L3, Ubiquitin-conjugating enzyme E2-F1, ubiquitin-conjugating enzyme E2L 3, ubiquitin-conjugating enzyme UBCH7, Ubiquitin-protein ligase L3
Gene Symbols UBE2L3
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human UBE2L3. The exact sequence is proprietary.
Molecular Weight of Antigen 18 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7332
Test Specificity Human, Mouse. Species reactivity based on sequence identity: Chicken (100%), Rhesus Monkey (100%), Pig (100%), Bovine (100%).
Target Species Human, Mouse
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.