Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UbcH7/UBE2L3 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB292461 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292461 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292461 Supplier Novus Biologicals Supplier No. NB1002265PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UbcH7/UBE2L3 Polyclonal specifically detects UbcH7/UBE2L3 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot.

Specifications

Antigen UbcH7/UBE2L3
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias E2-F1, EC 6.3.2.19, L-UBC, UBCE7, UbcH7, UBCH7UbcM4, Ubiquitin carrier protein L3, ubiquitin-conjugating enzyme E2 L3, Ubiquitin-conjugating enzyme E2-F1, ubiquitin-conjugating enzyme E2L 3, ubiquitin-conjugating enzyme UBCH7, Ubiquitin-protein ligase L3
Host Species Rabbit
Immunogen Maps to a region between residue 100 and the C-terminus (residue 154) of human Ubiquitin-Conjugating Enzyme H7 using the numbering given in entry NP_003338.1 (GeneID 7332).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cancer, DNA Repair, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 7332
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.