Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2D4/Ubch5d Antibody (129), PE, Novus Biologicals™
SDP

Catalog No. NB316536 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB316536 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB316536 Supplier Novus Biologicals Supplier No. NBP290194PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

UBE2D4/Ubch5d Monoclonal antibody specifically detects UBE2D4/Ubch5d in Human samples. It is validated for ELISA

Specifications

Antigen UBE2D4/Ubch5d
Applications ELISA
Classification Monoclonal
Clone 129
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias EC 6.3.2.19, FLJ32004, HBUCE1Ubiquitin carrier protein D4, UBCH5D, ubiquitin-conjugating enzyme E2 D4, ubiquitin-conjugating enzyme E2D 4 (putative), ubiquitin-conjugating enzyme HBUCE1, Ubiquitin-protein ligase D4
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human UBE2D4/Ubch5d (Uniprot#: Q9Y2X8; Met1-Met1474).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 51619
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.