Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2E3 Antibody (OTI7E8), Alexa Fluor™ 700, Novus Biologicals™
SDP

Catalog No. N274745A700 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274745A700 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274745A700 Supplier Novus Biologicals Supplier No. NBP274745AF700
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2E3 Monoclonal specifically detects UBE2E3 in Human, Mouse, Rat, Canine, Primate, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen UBE2E3
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI7E8
Conjugate Alexa Fluor 700
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2.19, UBCE4, UbcH9, UBCH9ubiquitin-conjugating enzyme E2E 3 (homologous to yeast UBC4/5), UbcM2, Ubiquitin carrier protein E3, ubiquitin-conjugating enzyme E2 E3, Ubiquitin-conjugating enzyme E2-23 kDa, ubiquitin-conjugating enzyme E2E 3 (UBC4/5 homolog, yeast), Ubiquitin-protein ligase E3
Gene Symbols UBE2E3
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2E3 (NP_006348) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 10477.0
Target Species Human, Mouse, Rat, Canine, Primate, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.