Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2F/NCE2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP219485 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP219485 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP219485 Supplier Novus Biologicals Supplier No. NBP219485
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2F/NCE2 Polyclonal specifically detects UBE2F/NCE2 in Human samples. It is validated for Western Blot.

Specifications

Antigen UBE2F/NCE2
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. Q969M7
Gene Alias EC 6.3.2, EC 6.3.2.-, MGC18120, NCE2Ubiquitin-conjugating enzyme E2 F, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme, NEDD8 protein ligase UBE2F, NEDD8-conjugating enzyme 2, NEDD8-conjugating enzyme UBE2F, ubiquitin-conjugating enzyme E2F (putative)
Gene Symbols UBE2F
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human NCE2. The exact sequence is proprietary.
Molecular Weight of Antigen 21 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140739
Test Specificity Human. Species reactivity based on sequence identity: Rhesus Monkey (100%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.