Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2F/NCE2 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB322677 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322677 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322677 Supplier Novus Biologicals Supplier No. NBP300049PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2F/NCE2 Polyclonal antibody specifically detects UBE2F/NCE2 in Human samples. It is validated for ELISA

Specifications

Antigen UBE2F/NCE2
Applications ELISA
Classification Polyclonal
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias EC 6.3.2, EC 6.3.2.-, MGC18120, NCE2Ubiquitin-conjugating enzyme E2 F, NEDD8 carrier protein UBE2F, NEDD8 conjugating enzyme, NEDD8 protein ligase UBE2F, NEDD8-conjugating enzyme 2, NEDD8-conjugating enzyme UBE2F, ubiquitin-conjugating enzyme E2F (putative)
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human UBE2F/NCE2 (Uniprot#: Q969M7-1; Met1-Arg185)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140739
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.