Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2G1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP213499 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213499 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213499 Supplier Novus Biologicals Supplier No. NBP213499
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2G1 Polyclonal specifically detects UBE2G1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UBE2G1
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 6.3.2.19, UBC7, UBE2GE217K, Ubiquitin carrier protein G1, ubiquitin-conjugating enzyme E2 G1, ubiquitin-conjugating enzyme E2G 1 (homologous to C. elegans UBC7), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, C. elegans), ubiquitin-conjugating enzyme E2G 1 (UBC7 homolog, yeast), Ubiquitin-protein ligase G1
Gene Symbols UBE2G1
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the amino acids: MTELQSALLLRRQLAELNKNPVEGFSAGLIDDNDLYRWEVLIIGPPDTLYE
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 7326
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.