Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2G2 Antibody, Novus Biologicals™
SDP

Catalog No. NB431621 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB431621 25 μL
NBP186888 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB431621 Supplier Novus Biologicals Supplier No. NBP18688825UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2G2 Polyclonal specifically detects UBE2G2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UBE2G2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Alias EC 6.3.2.19, UBC7human ubiquitin conjugating enzyme G210ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7), Ubiquitin carrier protein G2, ubiquitin conjugating enzyme 7, ubiquitin conjugating enzyme G2, ubiquitin-conjugating enzyme E2 G2, ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast), Ubiquitin-protein ligase G2
Gene Symbols UBE2G2
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids:LSFPLDYPLSPPKMRFTCEMFHPNIYPDGRVCISILHAPGDDPMGYESSAERWSPVQSVEKILLSVVSMLAEPNDESGANVDASKMWRDDREQFYKIAKQIVQKS
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7327
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.