Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2G2 Antibody (OTI1B10), Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. N274746A405 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274746A405 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274746A405 Supplier Novus Biologicals Supplier No. NBP274746AF405

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Mouse Monoclonal Antibody

UBE2G2 Monoclonal specifically detects UBE2G2 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UBE2G2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1B10
Conjugate Alexa Fluor 405
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 6.3.2.19, UBC7human ubiquitin conjugating enzyme G210ubiquitin-conjugating enzyme E2G 2 (homologous to yeast UBC7), Ubiquitin carrier protein G2, ubiquitin conjugating enzyme 7, ubiquitin conjugating enzyme G2, ubiquitin-conjugating enzyme E2 G2, ubiquitin-conjugating enzyme E2G 2 (UBC7 homolog, yeast), Ubiquitin-protein ligase G2
Gene Symbols UBE2G2
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2G2 (NP_003334) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 7327.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.