Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2I/Ubc9 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB322484 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB322484 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB322484 Supplier Novus Biologicals Supplier No. NBP300096MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2I/Ubc9 Polyclonal antibody specifically detects UBE2I/Ubc9 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen UBE2I/Ubc9
Applications Western Blot, Immunocytochemistry, Immunoprecipitation
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias C358B7.1, EC 6.3.2, EC 6.3.2.-, P18, SUMO-1-protein ligase, SUMO-protein ligase, UBC9SUMO-conjugating enzyme UBC9, UBCE9, Ubiquitin carrier protein 9, Ubiquitin carrier protein I, ubiquitin conjugating enzyme 9, Ubiquitin-conjugating enzyme E2 I, ubiquitin-conjugating enzyme E2I (homologous to yeast UBC9), ubiquitin-conjugating enzyme E2I (UBC9 homolog, yeast), ubiquitin-conjugating enzyme UbcE2A, ubiquitin-like protein SUMO-1 conjugating enzyme, ubiquitin-protein ligase E2I, Ubiquitin-protein ligase I
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant Human UBE2I/Ubc9 (Accession#: P63279; Met1-Ser158)
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 7329
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.