Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J1 Antibody (OTI3D1), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274747A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274747A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274747A94 Supplier Novus Biologicals Supplier No. NBP274747AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2J1 Monoclonal specifically detects UBE2J1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen UBE2J1
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone OTI3D1
Conjugate Alexa Fluor 594
Dilution Western Blot, Flow Cytometry, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias CGI-76, EC 6.3.2.19, HSPC153, HSPC205, HSU93243, HsUBC6e, MGC12555, NCUBE-1, NCUBE1HSUBC6e, Non-canonical ubiquitin-conjugating enzyme 1, non-canonical ubquitin conjugating enzyme 1, Ubc6p, ubiquitin-conjugating enzyme E2 J1, ubiquitin-conjugating enzyme E2, J1 (UBC6 homolog, yeast), Yeast ubiquitin-conjugating enzyme UBC6 homolog E
Gene Symbols UBE2J1
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2J1 (NP_057105) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 51465
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.