Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J2/UBC6 Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NB136277 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB136277 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB136277 Supplier Novus Biologicals Supplier No. NBP306347AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2J2/UBC6 Polyclonal specifically detects UBE2J2/UBC6 in Human samples. It is validated for Western Blot.

Specifications

Antigen UBE2J2/UBC6
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias EC 6.3.2.19, J2, Ubc6p, ubiquitin-conjugating enzyme E2 J2, ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast), yeast UBC6 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human UBE2J2/UBC6 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 118424
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.