Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2J2/UBC6 Antibody, Novus Biologicals™
SDP

Catalog No. NBP178071 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP178071 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP178071 Supplier Novus Biologicals Supplier No. NBP178071
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2J2/UBC6 Polyclonal specifically detects UBE2J2/UBC6 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunoprecipitation.

Specifications

Antigen UBE2J2/UBC6
Applications Western Blot, ELISA, Immunoprecipitation
Classification Polyclonal
Concentration 0.91 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:5000-1:25000, Immunoprecipitation
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q8N2K1
Gene Alias EC 6.3.2.19, J2, Ubc6p, ubiquitin-conjugating enzyme E2 J2, ubiquitin-conjugating enzyme E2, J2 (UBC6 homolog, yeast), yeast UBC6 homolog
Gene Symbols UBE2J2
Host Species Rabbit
Immunogen This affinity purified UBE2J2/UBC6 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region of the human UBE2J2/UBC6 protein. (Uniprot: Q8N2K1)
Molecular Weight of Antigen 28.9 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 118424
Test Specificity This affinity purified antibody is directed against human Ube2j2 protein. A BLAST analysis was used to suggest cross-reactivity with Ube2j2 protein from human
Target Species Human, Mouse, Rat, Avian, Bovine, Canine, Chicken, Equine, Fish, Mammalia, Monkey, Opossum, Xenopus species
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.