Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2M/Ubc12 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB318776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318776 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318776 Supplier Novus Biologicals Supplier No. NBP298833APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE2M/Ubc12 Polyclonal antibody specifically detects UBE2M/Ubc12 in Human samples. It is validated for Western Blot

Specifications

Antigen UBE2M/Ubc12
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias EC 6.3.2, EC 6.3.2.-, hUbc12, NEDD8 carrier protein, NEDD8 protein ligase, UBC12NEDD8-conjugating enzyme Ubc12, UBC-RS2, ubiquitin carrier protein M, Ubiquitin-conjugating enzyme E2 M, ubiquitin-conjugating enzyme E2M (homologous to yeast UBC12), ubiquitin-conjugating enzyme E2M (UBC12 homolog, yeast), ubiquitin-protein ligase M, yeast UBC12 homolog
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human UBE2M/Ubc12.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins
Primary or Secondary Primary
Gene ID (Entrez) 9040
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.