Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2O Antibody, Novus Biologicals™
SDP

Catalog No. NBP103336 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP103336 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP103336 Supplier Novus Biologicals Supplier No. NBP103336
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

UBE2O Polyclonal specifically detects UBE2O in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen UBE2O
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 mcg/mg lysate
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. Q9C0C9
Gene Alias E2-230K, EC 6.3.2.19, FLJ12878, KIAA1734, Ubiquitin carrier protein O, ubiquitin-conjugating enzyme E2 O, Ubiquitin-conjugating enzyme E2 of 230 kDa, Ubiquitin-conjugating enzyme E2-230K, ubiquitin-conjugating enzyme E2O, Ubiquitin-protein ligase O
Gene Symbols UBE2O
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 725 and 775 of human ubiquitin-conjugating enzyme E2O using the numbering given in entry NP_071349.2 (GeneID 63893).
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 63893
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, White-tufted-ear marmoset, Crab-eating macaque, European domestic ferret, African elephant, Small-eared galago and Little brown bat.
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.