Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2R1/CDC34 Antibody (CPTC-CDC34-2), DyLight 680, Novus Biologicals™
SDP

Catalog No. NB384140 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB384140 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB384140 Supplier Novus Biologicals Supplier No. NBP308839FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2R1/CDC34 Monoclonal antibody specifically detects UBE2R1/CDC34 in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen UBE2R1/CDC34
Applications Immunohistochemistry (Paraffin), Protein Array
Classification Monoclonal
Clone CPTC-CDC34-2
Conjugate DyLight 680
Formulation 50mM Sodium Borate
Gene Alias cell division cycle 34, cell division cycle 34 homolog (S. cerevisiae), E2-CDC34, EC 6.3.2.19, UBC3, UBE2R1UBCH3, ubiquitin carrier protein, ubiquitin-conjugating enzyme E2 R1, Ubiquitin-conjugating enzyme E2-32 kDa complementing, Ubiquitin-conjugating enzyme E2-CDC34, Ubiquitin-protein ligase R1
Host Species Mouse
Immunogen Recombinant human full-length UBE2R1/CDC34 protein (Uniprot: P49427)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication, Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 997
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.