Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2S Antibody (2C12), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP141126A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP141126A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP141126A70 Supplier Novus Biologicals Supplier No. NBP141126AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2S Monoclonal specifically detects UBE2S in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen UBE2S
Applications Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 2C12
Conjugate Alexa Fluor 750
Dilution Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S
Gene Symbols UBE2S
Host Species Mouse
Immunogen Recombinant human UBE2S (1-222aa) purified from E. coli (NP_055316.2).
Purification Method Protein G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27338.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.