Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE2S Antibody (OTI1B5), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274749A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274749A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274749A30 Supplier Novus Biologicals Supplier No. NBP274749AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBE2S Monoclonal specifically detects UBE2S in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.

Specifications

Antigen UBE2S
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone OTI1B5
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias E2-EPF5, E2-EPFEC 6.3.2.19, E2EPFEPF5, Ubiquitin carrier protein S, ubiquitin-conjugating enzyme E2 S, ubiquitin-conjugating enzyme E2-24 kD, Ubiquitin-conjugating enzyme E2-24 kDa, Ubiquitin-conjugating enzyme E2-EPF5, ubiquitin-conjugating enzyme E2S, Ubiquitin-protein ligase S
Gene Symbols UBE2S
Host Species Mouse
Immunogen Full length recombinant protein of human UBE2S (NP_055316) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 27338
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.