Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Recombinant Human UBE2Z His Protein
SDP

Catalog No. p-7104637 Shop All R&D Systems Products
Click to view available options
Quantity:
0.05 mg
0.25 mg

Catalog No. NBP25173825

Supplier: Novus Biologicals™ NBP2517380.25MG

Only null left
Add to Cart
Add to Cart

Highly purified. Generating reliable and reproducible results.

A recombinant protein with a N-Terminal His-tag and corresponding to the amino acids 1-246 of Human UBE2Z The Recombinant Human UBE2Z Protein is derived from E. coli. The Recombinant Human UBE2Z Protein has been validated for the following applications: SDS-Page.

Specifications

Concentration 0.25mg/mL
For Use With (Application) SDS-PAGE
Formulation 20mM Tris-HCl buffer (pH 8.0) containing 0.15M NaCl, 20% glycerol, 1mM DTT
Gene ID (Entrez) 65264
Molecular Weight (g/mol) TMW: 30.5kDa
Quantity 0.25 mg
Storage Requirements Store at 4°C short term. Aliquot and store at −20°C long term. Avoid freeze-thaw cycles.
Gene Alias EC 6.3.2.19, FLJ13855, Uba6-specific E2 conjugating enzyme 1, UBA6-specific enzyme E2, Ubiquitin carrier protein Z, ubiquitin-conjugating enzyme E2 Z, ubiquitin-conjugating enzyme E2Z, Ubiquitin-protein ligase Z, USE1
Common Name UBE2Z
Conjugate Unconjugated
Recombinant Recombinant
Purity or Quality Grade >95%, by SDS-PAGE under reducing conditions and visualized by Colloidal Coomassie™ Blue stain
Show More Show Less

For research use only and is not approved for use in humans or in clinical diagnosis

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.