Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBE4B Antibody, DyLight 405, Novus Biologicals™
SDP

Catalog No. NB056790 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB056790 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB056790 Supplier Novus Biologicals Supplier No. NBP298954V
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBE4B Polyclonal specifically detects UBE4B in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen UBE4B
Applications Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 405
Dilution Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias E4, homologous to yeast UFD2, Homozygously deleted in neuroblastoma 1, homozygously deleted in neuroblastoma-1, KIAA0684HDNB1, ubiquitin conjugation factor E4 B, Ubiquitin fusion degradation protein 2, ubiquitination factor E4B (homologous to yeast UFD2), ubiquitination factor E4B (UFD2 homolog, yeast), ubiquitin-fusion degradation protein 2, UBOX3, UFD2UFD2A
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human UBE4B.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 10277
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.