Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBIAD1 Antibody, mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB341367 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341367 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341367 Supplier Novus Biologicals Supplier No. NBP282046MFV450
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBIAD1 Polyclonal antibody specifically detects UBIAD1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen UBIAD1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 450 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EC 2.5.1.-, RP4-796F18.1, SCCD, Schnyder crystalline corneal dystrophy, TERE1transitional epithelia response protein, Transitional epithelial response protein 1, UbiA prenyltransferase domain containing 1, ubiA prenyltransferase domain-containing protein 1
Host Species Rabbit
Immunogen UBIAD1 antibody was raised against a 15 amino acid peptide near the carboxy terminus of human UBIAD1. The immunogen is located within amino acids 210 - 260 of UBIAD1. Amino Acid Squence: LHSNNTRDMESDREA
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 29914
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.