Learn More
Description
Specifications
Specifications
| Antigen | Ubiquilin 2 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown |
| Classification | Monoclonal |
| Clone | 6H9 |
| Conjugate | Alexa Fluor 750 |
| Dilution | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated |
| Formulation | 50mM Sodium Borate with 0.05% Sodium Azide |
| Gene Alias | Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2 |
| Gene Symbols | UBQLN2 |
| Host Species | Mouse |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
