Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquilin 2 Antibody (6H9), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB005333 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005333 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005333 Supplier Novus Biologicals Supplier No. NBP225164JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquilin 2 Monoclonal specifically detects Ubiquilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Knockdown Validated.

Specifications

Antigen Ubiquilin 2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Classification Monoclonal
Clone 6H9
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, KnockDown Validated
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2
Gene Symbols UBQLN2
Host Species Mouse
Immunogen Human Ubiquilin 2 expressed in and purified from E. coli [UniProt# Q9UHD9]
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.