Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquilin 2 Antibody (6H9), Novus Biologicals™
SDP

Catalog No. NBP225164 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP225164 0.1 mL
NBP225164SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP225164 Supplier Novus Biologicals Supplier No. NBP225164
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 8 publications

Ubiquilin 2 Monoclonal specifically detects Ubiquilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockdown Validated.

Specifications

Antigen Ubiquilin 2
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay, KnockDown
Classification Monoclonal
Clone 6H9
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. Q9UHD9
Gene Alias Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2
Gene Symbols UBQLN2
Host Species Mouse
Immunogen Human Ubiquilin 2 expressed in and purified from E. coli [UniProt# Q9UHD9]
Molecular Weight of Antigen 66-68 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29978
Test Specificity Human and Mouse. Other species have not been tested.
Target Species Human, Mouse, Rat
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.