Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquilin 2 Antibody (6H9), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 8 publications
$243.50 - $489.50
Specifications
| Antigen | Ubiquilin 2 |
|---|---|
| Clone | 6H9 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000-1:2000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay, KnockDown |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP225164SS
![]() |
Novus Biologicals
NBP225164SS |
0.025 mL |
Each for $243.50
|
|
|||||
NBP225164
![]() |
Novus Biologicals
NBP225164 |
0.1 mL |
Each for $489.50
|
|
|||||
Description
Ubiquilin 2 Monoclonal specifically detects Ubiquilin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockdown Validated.Specifications
| Ubiquilin 2 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Frozen), Immunoassay, KnockDown | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| Q9UHD9 | |
| 29978 | |
| Human Ubiquilin 2 expressed in and purified from E. coli [UniProt# Q9UHD9] | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| 66-68 kDa |
| 6H9 | |
| Western Blot 1:1000-1:2000, Simple Western 1:100, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000, Immunoprecipitation, Immunohistochemistry-Frozen, Knockout Validated, Knockdown Validated | |
| Monoclonal | |
| Purified | |
| RUO | |
| 50% PBS, 50% glycerol with 5mM Sodium Azide | |
| Chap1, CHAP1/DSK2, DSK2, DSK2 homolog, FLJ10167, FLJ56541, hPLIC-2, N4BP4LIC-2, Nedd4 binding protein 4, PLIC-2, PLIC2Dsk2, Protein linking IAP with cytoskeleton 2, RIHFB2157, ubiquilin 2, ubiquilin-2, Ubiquitin-like product Chap1/Dsk2 | |
| UBQLN2 | |
| IgG1 | |
| Protein G purified | |
| Human and Mouse. Other species have not been tested. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title