Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Antibody (1B4-UB) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB600776 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600776 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600776 Supplier Novus Biologicals Supplier No. NB600776
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Bovine samples. It is validated for Western Blot, Simple Western, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Ubiquitin
Applications Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 1B4-UB
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:1000, Simple Western, Flow Cytometry 1:10-1:50, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:500-1:1000, Immunohistochemistry-Frozen 1:500-1:1000
Gene Accession No. P0CG53
Gene Alias RPS27A, UBA52, UBB ubiquitin B, UBC
Gene Symbols RPS27A
Host Species Mouse
Immunogen Ubiquitin from bovine erythrocytes conjugated to bovine gamma globulin
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity Reacts with Bovine. Cross reacts with Human.
Target Species Human, Bovine
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.