Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Antibody (P4G7-H11), Novus Biologicals™
SDP

Catalog No. NBP119306 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.05 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP119306 0.05 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP119306 Supplier Novus Biologicals Supplier No. NBP119306
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Mouse, Rat, Bovine, Hamster, Rabbit samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry.

Specifications

Antigen Ubiquitin
Applications Western Blot, Flow Cytometry, Immunohistochemistry
Classification Monoclonal
Clone P4G7-H11
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Flow Cytometry 10 ug/ml, Immunohistochemistry
Formulation PBS containing 50% glycerol with 0.09% Sodium Azide
Gene Alias RPS27A, UBA52, UBB ubiquitin B, UBC
Gene Symbols RPS27A
Host Species Mouse
Immunogen Native bovine Ubiquitin
Purification Method Protein G purified
Quantity 0.05 mg
Regulatory Status RUO
Research Discipline Alzheimers Research, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity Ubiquitin (P4G7-H11)
Target Species Human, Mouse, Rat, Bovine, Hamster, Rabbit
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.