Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Antibody (Ubi-1), FITC, Novus Biologicals™
SDP

Catalog No. NB362843 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362843 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362843 Supplier Novus Biologicals Supplier No. NB300130F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot

Specifications

Antigen Ubiquitin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone Ubi-1
Conjugate FITC
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation PBS
Gene Alias RPS27A, UBA52, UBB ubiquitin B, UBC
Host Species Mouse
Immunogen Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Alzheimers Research, Autophagy, Cancer, Membrane Trafficking and Chaperones, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Protein Turnover, Ubiquitin Proteasome Pathway
Primary or Secondary Primary
Gene ID (Entrez) 6233
Target Species Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.