Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Antibody (Ubi-1), Novus Biologicals™
SDP

Catalog No. NB300130SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB300130SS 0.025 mL
NB300130 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB300130SS Supplier Novus Biologicals Supplier No. NB300130SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody has been used in 32 publications

Ubiquitin Monoclonal specifically detects Ubiquitin in Human, Mouse, Rat, Porcine, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.

Specifications

Antigen Ubiquitin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone Ubi-1
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:2000, Immunohistochemistry 1:2000, Immunocytochemistry/Immunofluorescence 1:2000, Immunohistochemistry-Paraffin 1:2000, Immunohistochemistry-Frozen 1:2000, Immunoblotting
Formulation 50% PBS, 50% glycerol with 5mM Sodium Azide
Gene Accession No. P0CG48, P0CG47
Gene Alias RPS27A, UBA52, UBB ubiquitin B, UBC
Gene Symbols RPS27A
Host Species Mouse
Immunogen Purified bovine erythrocyte ubiquitin cross-linked with glutaraldehyde to KLH
Molecular Weight of Antigen 8.5 kDa
Purification Method Affinity purified
Quantity 0.025 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6233
Test Specificity Recognizes polyubiquitin chains much more strongly than monoubiquitinated molecules or free ubiquitin. Specifically recognizes ubiquitinated cytoplasmic inclusion bodies.
Target Species Human, Mouse, Rat, Pig, Bovine, C. elegans, Chicken, Drosophila, Equine, Plant, Monkey, Zebrafish
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Ascites
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.