Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin B Antibody (6C11-B3), FITC, Novus Biologicals™
SDP

Catalog No. NBP212882F Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP212882F 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP212882F Supplier Novus Biologicals Supplier No. NBP212882F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquitin B Monoclonal specifically detects Ubiquitin B in Human, Mouse, Rat, Bovine samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Ubiquitin B
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone 6C11-B3
Conjugate FITC
Dilution ELISA, Immunocytochemistry/Immunofluorescence
Formulation PBS with 0.05% Sodium Azide
Gene Alias FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B
Gene Symbols UBB
Host Species Mouse
Immunogen Native bovine ubiquitin, conjugated to KLH
Purification Method Protein G purified
Quantity 200 μL
Primary or Secondary Primary
Gene ID (Entrez) 7314
Test Specificity Detects approx 10kDa.
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.