Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin B Antibody (UBB/1748), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB389555 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB389555 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB389555 Supplier Novus Biologicals Supplier No. NBP308905DL594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ubiquitin B Monoclonal antibody specifically detects Ubiquitin B in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein Array

Specifications

Antigen Ubiquitin B
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone UBB/1748
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias FLJ25987, MGC8385, polyubiquitin B, polyubiquitin-B, RPS27A, UBA52, UBC, ubiquitin B
Host Species Mouse
Immunogen Recombinant fragment of human Ubiquitin B protein (around aa 1-119) (exact sequence is proprietary)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Alzheimers Research, Apoptosis, Cell Biology, Neurodegeneration, Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 7314
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2c κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.