Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ubiquitin Mouse anti-Human, Clone: Ubi-1, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1510I100UG
Description
Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications
Ubiquitin | |
Monoclonal | |
1.0 mg/mL | |
Purified mouse monoclonal antibody IgG1 in buffer containing PBS with 5 mM sodium azide with 50% glycerol. | |
Purified ubiquitin from bovine red blood cells covalently coupled to KLH using glutaraldehyde. | |
100 μg | |
Primary | |
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance. | |
IgG1 κ |
Immunohistochemistry, Western Blot | |
Ubi-1 | |
Unconjugated | |
Mouse | |
Protein G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction