Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ubiquitin Mouse anti-Human, Clone: Ubi-1, MilliporeSigma™

Mouse Monoclonal Antibody

$188.90 - $486.00

Specifications

Antigen Ubiquitin
Clone Ubi-1
Concentration 1.0 mg/mL
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MAB1510I25
View Documents
MilliporeSigma
MAB1510I25UG
25 μg
Each for $188.90
Only null left
Add to Cart
 
MAB1510I100
View Documents
MilliporeSigma
MAB1510I100UG
100 μg
Each for $486.00
Only null left
Add to Cart
 
Description

Description

Ubiquitin Monoclonal antibody specifically detects Ubiquitin in Human samples. It is validated for Immunohistochemistry, Western Blotting.
Specifications

Specifications

Ubiquitin
1.0 mg/mL
Monoclonal
Purified
RUO
Purified mouse monoclonal antibody IgG1 in buffer containing PBS with 5 mM sodium azide with 50% glycerol.
IgG1 κ
Protein G purified
Ubi-1
Immunohistochemistry, Western Blot
Unconjugated
Mouse
Human
Purified ubiquitin from bovine red blood cells covalently coupled to KLH using glutaraldehyde.
Primary
Stable for 1 year at -10°C to -25°C from date of receipt. Handling Recommendations: Upon receipt and prior to removing the cap, centrifuge the vial and gently mix the solution. Aliquot into microcentrifuge tubes and store at -20°C. Avoid repeated freeze/thaw cycles, which may damage IgG and affect product performance.
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.