Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBL3 Antibody (OTI1G4), Biotin, Novus Biologicals™
SDP

Catalog No. NBP274751B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274751B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274751B Supplier Novus Biologicals Supplier No. NBP274751B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBL3 Monoclonal specifically detects UBL3 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen UBL3
Applications Western Blot
Classification Monoclonal
Clone OTI1G4
Conjugate Biotin
Dilution Western Blot
Formulation PBS with 0.05% Sodium Azide
Gene Alias DKFZP434K151, FLJ32018, HCG-1, hsMUB, Membrane-anchored ubiquitin-fold protein, MUB, PNSC1, Protein HCG-1, ubiquitin-like 3, ubiquitin-like protein 3
Gene Symbols UBL3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-114 of human UBL3 (NP_009037) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 5412.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.