Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBL5 Antibody, Novus Biologicals™
SDP

Catalog No. NB600470 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB600470 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB600470 Supplier Novus Biologicals Supplier No. NB600470
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

UBL5 Polyclonal specifically detects UBL5 in Yeast samples. It is validated for Western Blot, ELISA.

Specifications

Antigen UBL5
Applications Western Blot, ELISA
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, ELISA 1:1000-1:5000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. Q6Q546
Gene Alias FLJ46917, HUB1, MGC131795, ubiquitin-like 5, ubiquitin-like protein 5
Gene Symbols UBL5
Host Species Rabbit
Immunogen This purified UBL5 Antibody was prepared from rabbit serum after repeated immunizations with recombinant yeast UBL5 protein. (Uniprot: Q6Q546)
Molecular Weight of Antigen 8.27 kDa
Purification Method Multi-step
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Neuroscience, Vision
Primary or Secondary Primary
Gene ID (Entrez) 59286
Test Specificity Hub 1
Reconstitution Reconstitute with 100 ul deionized water (or equivalent)
Target Species Yeast
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.