Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBN1 Antibody (UBN1-02), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NB002385 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB002385 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB002385 Supplier Novus Biologicals Supplier No. NBP262205AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBN1 Monoclonal specifically detects UBN1 in Human samples. It is validated for Western Blot.

Specifications

Antigen UBN1
Applications Western Blot
Classification Monoclonal
Clone UBN1-02
Conjugate Alexa Fluor 750
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4
Gene Symbols UBN1
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 29855
Test Specificity The antibody SC-05 reacts with 80 kDa human secretory component glycoprotein (both free and bound in secretory IgA). Recognized glycoprotein is specific membrane marker of glandular carcinomas.
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.