Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBN1 Antibody (UBN1-02), FITC, Novus Biologicals™
SDP

Catalog No. NB343086 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343086 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343086 Supplier Novus Biologicals Supplier No. NBP262205F
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBN1 Monoclonal antibody specifically detects UBN1 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen UBN1
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone UBN1-02
Conjugate FITC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias HIRA-binding protein, ubinuclein 1, ubinuclein-1, VT, VT4
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29855
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.