Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBOX5 Antibody (OTI1C5), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB274760APC Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB274760APC 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB274760APC Supplier Novus Biologicals Supplier No. NBP274760APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBOX5 Monoclonal specifically detects UBOX5 in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen UBOX5
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C5
Conjugate APC
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias KIAA0860U-box domain-containing protein 5, RING finger protein 37, RNF37Ubce7ip5, UBCE7IP5, ubiquitin conjugating enzyme 7 interacting protein 5, Ubiquitin-conjugating enzyme 7-interacting protein 5, U-box domain containing 5, UIP5
Gene Symbols UBOX5
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-130 and C 419-487 of human UBOX5 (NP_955447) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 22888.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.