Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBP1 Antibody (OTI9G7), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP274761A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274761A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274761A94 Supplier Novus Biologicals Supplier No. NBP274761AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

UBP1 Monoclonal specifically detects UBP1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.

Specifications

Antigen UBP1
Applications Immunohistochemistry
Classification Monoclonal
Clone OTI9G7
Conjugate Alexa Fluor 594
Dilution Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias DKFZp686L1745, LBP1, LBP-1a, LBP1A, LBP1B, LBP-1B, Transcription factor LBP-1, upstream binding protein 1 (LBP-1a), upstream-binding protein 1
Gene Symbols UBP1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 47-269 of human UBP1 (NP_055332) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7342.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.