Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

UBR2 Antibody (PCRP-UBR2-1D12), DyLight 405, Novus Biologicals™
SDP

Catalog No. NB157969 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB157969 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB157969 Supplier Novus Biologicals Supplier No. NBP314287V
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

UBR2 Monoclonal antibody specifically detects UBR2 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen UBR2
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-UBR2-1D12
Conjugate DyLight 405
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias bA49A4.1, C6orf133MGC71112, chromosome 6 open reading frame 133, dJ242G1.1, dJ392M17.3, DKFZp686C08114, E3 ubiquitin-protein ligase UBR2, EC 6.3.2.-, KIAA0349FLJ36357, N-recognin-2, RP3-392M17.3, ubiquitin protein ligase E3 component n-recognin 2, Ubiquitin-protein ligase E3-alpha-2, Ubiquitin-protein ligase E3-alpha-II
Host Species Mouse
Immunogen Recombinant full-length human UBR2 protein (Uniprot: Q8IWV8)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23304
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.